Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034267.2 | NP_001029439.1 | 3 - 4 | 403 | 111 | |
GenBank mRNA | BC102501.1 | - | 3 - 4 | 404 | 111 | |
BC142371.1 | - | 4 - 5 | 715 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |