Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034272.1 | NP_001029444.1 | 1 - 2 | 152 | 80 | |
GenBank mRNA | BC120382.1 | - | 1 - 2 | 154 | 80 | |
BT021507.1 | - | 1 - 2 | 152 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |