Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034319.2 | NP_001029491.1 | 7 - 8 | 720 | 73 | |
GenBank mRNA | BC102927.1 | - | 7 - 8 | 721 | 73 | |
EF576938.1 | - | 7 - 8 | 705 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |