Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034534.1 | NP_001029706.1 | 5 - 6 | 848 | 66 | |
GenBank mRNA | BC103222.1 | - | 5 - 6 | 848 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |