Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034543.1 | NP_001029715.1 | 1 - 2 | 120 | 139 | |
GenBank mRNA | BC102241.1 | - | 1 - 2 | 120 | 139 | |
BT020972.1 | - | 1 - 2 | 81 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |