Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034574.1 | NP_001029746.1 | 6 - 7 | 984 | 73 | |
GenBank mRNA | BC104589.1 | - | 6 - 7 | 1006 | 73 | |
BT021589.1 | - | 6 - 7 | 984 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |