Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034636.2 | NP_001029808.1 | 2 - 3 | 307 | 116 | |
GenBank mRNA | BC102885.1 | - | 2 - 3 | 309 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |