Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034643.2 | NP_001029815.1 | 3 - 4 | 464 | 75 | |
GenBank mRNA | BC102576.1 | - | 3 - 4 | 465 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |