Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034781.2 | NP_001029953.1 | 3 - 4 | 275 | 145 | |
GenBank mRNA | BC102435.1 | - | 3 - 4 | 278 | 145 | |
BT029901.1 | - | 3 - 4 | 253 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |