Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034811.1 | NP_001029983.1 | 2 - 3 | 425 | 74 | |
GenBank mRNA | BC103476.1 | - | 2 - 3 | 240 | 74 | |
BT021724.1 | - | 2 - 3 | 425 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |