Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035070.2 | NP_001030242.2 | 7 - 8 | 925 | 70 | |
GenBank mRNA | BC148874.1 | - | 7 - 8 | 926 | 70 | |
BT021867.1 | - | 2 - 3 | 354 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |