Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035090.1 | NP_001030262.1 | 6 - 7 | 721 | 148 | |
GenBank mRNA | BT021570.1 | - | 6 - 7 | 721 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |