Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035301.1 | NP_001030378.1 | 3 - 4 | 634 | 64 | |
GenBank mRNA | BC113220.1 | - | 3 - 4 | 582 | 64 | |
BT021182.1 | - | 3 - 4 | 634 | 64 | ||
BT026299.1 | - | 3 - 4 | 501 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |