Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035437.1 | NP_001030514.1 | 2 - 3 | 97 | 61 | |
GenBank mRNA | BC102229.1 | - | 2 - 3 | 97 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |