Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037320.1 | NP_001032397.1 | 5 - 6 | 604 | 121 | |
GenBank mRNA | BC122783.1 | - | 5 - 6 | 622 | 121 | |
BT021766.1 | - | 5 - 6 | 604 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |