Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040474.2 | NP_001035564.1 | 4 - 5 | 607 | 74 | |
GenBank mRNA | BC103099.1 | - | 4 - 5 | 609 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |