Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040543.3 | NP_001035633.1 | 2 - 3 | 143 | 72 | |
GenBank mRNA | BC103236.1 | - | 1 | 130 | 72 | |
CB426020.1 | - | 2 - 3 | 111 | 72 | ||
EV659942.1 | - | 2 - 3 | 147 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |