Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040593.2 | NP_001035683.1 | 3 - 4 | 350 | 58 | |
GenBank mRNA | AY911338.1 | - | 3 - 4 | 351 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |