Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040601.2 | NP_001035691.1 | 8 - 9 | 846 | 56 | |
GenBank mRNA | BC102118.1 | - | 8 - 9 | 847 | 56 | |
BT021000.1 | - | 8 - 9 | 861 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |