Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040604.1 | NP_001035694.1 | 3 - 4 | 575 | 55 | |
GenBank mRNA | BC113347.1 | - | 3 - 4 | 589 | 55 | |
BT021015.1 | - | 3 - 4 | 575 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |