Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076295.2 | NP_001069763.2 | 4 - 5 | 469 | 90 | |
GenBank mRNA | BC102911.1 | - | 4 - 5 | 470 | 90 | |
EH156727.1 | - | 4 - 5 | 465 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |