Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077040.1 | NP_001070508.1 | 2 - 3 | 286 | 67 | |
GenBank mRNA | BT021572.1 | - | 2 - 3 | 286 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |