Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173886.2 | NP_776311.2 | 2 - 3 | 288 | 77 | |
GenBank mRNA | EV607234.1 | - | 2 - 3 | 295 | 77 | |
EV611026.1 | - | 2 - 3 | 292 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |