Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173950.2 | NP_776375.1 | 5 - 6 | 728 | 70 | |
GenBank mRNA | AB004272.1 | - | 5 - 6 | 728 | 70 | |
BC112622.1 | - | 5 - 6 | 762 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |