Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173958.1 | NP_776383.1 | 2 - 3 | 784 | 64 | |
GenBank mRNA | AY034785.1 | - | 2 - 3 | 784 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |