Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174011.3 | NP_776436.1 | 7 - 8 | 623 | 124 | |
GenBank mRNA | BC102243.1 | - | 7 - 8 | 623 | 124 | |
U25687.1 | - | 6 - 7 | 535 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |