Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159412.1 | NP_001152884.1 | 7 - 8 | 664 | 57 | |
NM_001159413.1 | NP_001152885.1 | 7 - 8 | 645 | 57 | ||
NM_174078.3 | NP_776503.1 | 7 - 8 | 761 | 57 | ||
GenBank mRNA | BC102478.1 | - | 7 - 8 | 761 | 57 | |
DN283346.1 | - | 7 - 8 | 646 | 57 | ||
X67029.1 | - | 645 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |