Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316338.1 | NP_001303267.1 | 3 - 4 | 395 | 106 | |
NM_174100.2 | NP_776525.2 | 3 - 4 | 395 | 106 | ||
GenBank mRNA | AJ401268.1 | - | 3 - 4 | 359 | 106 | |
BC102217.1 | - | 3 - 4 | 326 | 106 | ||
BC142006.1 | - | 3 - 4 | 396 | 106 | ||
BC151427.1 | - | 3 - 4 | 326 | 106 | ||
BT021009.1 | - | 3 - 4 | 363 | 106 | ||
DT720873.1 | - | 3 - 4 | 336 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |