Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174117.1 | NP_776542.1 | 10 - 11 | 1075 | 97 | |
GenBank mRNA | AB059399.2 | - | 10 - 11 | 1075 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |