Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174117.1 | NP_776542.1 | 5 - 6 | 602 | 77 | |
GenBank mRNA | AB059399.2 | - | 5 - 6 | 602 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |