Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174251.1 | NP_776676.1 | 3 - 4 | 385 | 61 | |
GenBank mRNA | BC102269.1 | - | 3 - 4 | 393 | 61 | |
L35045.1 | - | 3 - 4 | 385 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |