Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174256.3 | NP_776681.1 | 5 - 6 | 863 | 93 | |
GenBank mRNA | AY048883.1 | - | 5 - 6 | 864 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Bos taurus Probe spans exons |