Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174263.2 | NP_776688.1 | 3 - 4 | 310 | 97 | |
GenBank mRNA | AY061959.1 | - | 3 - 4 | 310 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |