Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174274.1 | NP_776699.1 | 2 - 3 | 1557 | 108 | |
GenBank mRNA | AY027881.1 | - | 2 - 3 | 1596 | 108 | |
AY027882.1 | - | 2 - 3 | 1596 | 108 | ||
BC151599.1 | - | 2 - 3 | 1505 | 108 | ||
BT025444.1 | - | 2 - 3 | 1551 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |