Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174359.1 | NP_776784.1 | 3 - 4 | 511 | 70 | |
GenBank mRNA | BC134467.1 | - | 3 - 4 | 513 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |