Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174471.3 | NP_776896.1 | 1 - 2 | 79 | 55 | |
GenBank mRNA | AY295346.1 | - | 1 - 2 | 79 | 55 | |
BC102489.1 | - | 1 - 2 | 45 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |