Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174486.3 | NP_776911.2 | 7 - 8 | 705 | 132 | |
GenBank mRNA | AF268465.1 | - | 6 - 7 | 587 | 132 | |
AJ288914.2 | - | 6 - 7 | 587 | 132 | ||
BC102904.1 | - | 7 - 8 | 751 | 132 | ||
BT021018.1 | - | 7 - 8 | 705 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |