Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174490.3 | NP_776915.1 | 1 - 2 | 250 | 152 | |
GenBank mRNA | BC103271.1 | - | 1 - 2 | 250 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |