Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174535.2 | NP_776960.1 | 6 - 7 | 1216 | 103 | |
GenBank mRNA | AB060107.1 | - | 1 | 1091 | 103 | |
AB099079.1 | - | 1 - 2 | 238 | 103 | ||
AF013213.1 | - | 3 - 4 | 601 | 103 | ||
AJ238405.1 | - | 5 - 6 | 1028 | 103 | ||
BC105315.1 | - | 1 | 1094 | 103 | ||
BC142302.1 | - | 1 | 1096 | 103 | ||
BC151690.1 | - | 1 | 1095 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |