Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174601.2 | NP_777026.1 | 5 - 6 | 1668 | 107 | |
GenBank mRNA | AY039236.1 | - | 5 - 6 | 1668 | 107 | |
BC123803.1 | - | 5 - 6 | 1257 | 107 | ||
LC005614.1 | - | 5 - 6 | 1053 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |