Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 10 - 11 | 1601 | 80 | |
GenBank mRNA | AJ508718.1 | - | 10 - 11 | 1601 | 80 | |
AY052775.1 | - | 11 - 12 | 1685 | 80 | ||
BC133437.1 | - | 10 - 11 | 1646 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |