Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 1 - 2 | 224 | 109 | |
GenBank mRNA | AJ508718.1 | - | 1 - 2 | 224 | 109 | |
AY052775.1 | - | 2 - 3 | 308 | 109 | ||
BC133437.1 | - | 1 - 2 | 269 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |