Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 2 - 3 | 363 | 141 | |
GenBank mRNA | AJ508718.1 | - | 2 - 3 | 363 | 141 | |
AY052775.1 | - | 3 - 4 | 447 | 141 | ||
BC133437.1 | - | 2 - 3 | 408 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |