Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174734.2 | NP_777159.1 | 3 - 4 | 722 | 142 | |
GenBank mRNA | AF456766.1 | - | 3 - 4 | 722 | 142 | |
BC134557.1 | - | 3 - 4 | 663 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |