Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174740.2 | NP_777165.1 | 4 - 5 | 657 | 66 | |
GenBank mRNA | AF490399.1 | - | 4 - 5 | 657 | 66 | |
AY075102.1 | - | 4 - 5 | 657 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |