Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174752.3 | NP_777177.1 | 3 - 4 | 101 | 120 | |
GenBank mRNA | AY154893.1 | - | 3 - 4 | 101 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |