Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174773.4 | NP_777198.2 | 7 - 8 | 1043 | 100 | |
GenBank mRNA | AF120106.1 | - | 6 - 7 | 968 | 100 | |
BC102907.1 | - | 7 - 8 | 1044 | 100 | ||
BT020785.1 | - | 7 - 8 | 1026 | 100 | ||
BT021173.1 | - | 7 - 8 | 1029 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |