Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174831.2 | NP_777256.1 | 3 - 4 | 405 | 79 | |
GenBank mRNA | BC149488.1 | - | 3 - 4 | 405 | 79 | |
BC149577.1 | - | 3 - 4 | 405 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |