Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175714.2 | NP_783645.1 | 2 - 3 | 310 | 67 | |
GenBank mRNA | AB100737.1 | - | 2 - 3 | 310 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |