Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175788.1 | NP_786982.1 | 5 - 6 | 1198 | 91 | |
GenBank mRNA | Y11045.1 | - | 1230 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |